missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Ship2 (Tyr1135) Recombinant Rabbit Monoclonal Antibody (Ship2Y1135-1D2), FITC, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Thermo Scientific MA537308
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Phospho-Ship2 (Tyr1135) Recombinant Monoclonal antibody specifically detects Phospho-Ship2 (Tyr1135) in Human, Mouse samples. It is validated for Flow Cytometry.Specifica
Phospho-Ship2 (Tyr1135) | |
Monoclonal | |
FITC | |
INPPL1 | |
51C; 51C protein; ablSH3-binding protein; inositol polyphosphate phosphatase like 1; inositol polyphosphate phosphatase-like 1; inositol polyphosphate phosphatase-like protein 1; Inppl1; INPPL-1; OPSMD; Phosphatidylinositol 3,4,5-trisphosphate 5-phosphatase 2; phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase 2; protein 51C; SH2 domain-containing inositol 5'-phosphatase 2; SH2 domain-containing inositol phosphatase 2; SH2 domain-containing inositol-5'-phosphatase 2; SH2-containing inositol phosphatase 2; SHIP2; SHIP-2 | |
Rabbit | |
Protein A/G | |
RUO | |
16332, 3636 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
Ship2Y1135-1D2 | |
PBS with 0.20% BSA and 0.09% sodium azide | |
O15357, Q6P549 | |
INPPL1 | |
A synthetic phospho-peptide corresponding to residues surrounding Tyr1135 of human phospho Ship2. | |
10 Tests | |
Primary | |
Human, Mouse | |
Antibody | |
IgG κ |