missing translation for 'onlineSavingsMsg'
Learn More

Phospho-TGFBR1 (Thr204) Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA564809

Dettagli aggiuntivi : Peso : 0.01000kg

Codice prodotto. 15712876

  • 463.00€ / 100µL
In stock
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Phospho-TGFBR1 (Thr204) Polyclonal Antibody detects endogenous levels of TGFBR1 only when phosphorylated at Thr204.

Phospho-TGFBR1 (Thr204) Polyclonal antibody specifically detects Phospho-TGFBR1 (Thr204) in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifica

Specifica

Phospho-TGFBR1 (Thr204)
Polyclonal
Unconjugated
TGFBR1
AAT5; activin A receptor type II-like kinase, 53kDa; activin A receptor type II-like protein kinase of 53kD; Activin receptor-like kinase 5; ACVRLK4; ALK5; Alk-5; AU017191; ESK2; ESS1; LDS1; LDS1A; LDS2A; MSSE; mutant transforming growth factor beta receptor I; ser; serine/threonine-protein kinase receptor R4; SKR4; TbetaRI; tbetaR-I; tgf beta receptor 1; TGF-beta receptor type I; TGF-beta receptor type-1; TGF-beta type I receptor; TGFBR1; TGFR1; TGFR-1; Transfor; transforming growth factor beta receptor 1; transforming growth factor beta receptor I; transforming growth factor, beta receptor 1; transforming growth factor, beta receptor I; transforming growth factor, beta receptor I (activin A receptor type II-like kinase, 53kDa); transforming growth factor-beta receptor type I
Rabbit
Affinity chromatography
RUO
21812, 29591, 7046
-20°C
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P36897, P80204, Q64729
TGFBR1
A synthesized peptide derived from human TGFBR1 (Phospho-Thr204)
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali