missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-ZAP70/Syk (Tyr319, Tyr352) Recombinant Rabbit Monoclonal Antibody (Zap70Y319-A3), APC, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Thermo Scientific MA536964
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Phospho-ZAP70/Syk (Tyr319, Tyr352) Recombinant Monoclonal antibody specifically detects Phospho-ZAP70/Syk (Tyr319, Tyr352) in Human, Mouse samples. It is validated for Flow Cytometry.Specifica
Phospho-ZAP70/Syk (Tyr319, Tyr352) | |
Monoclonal | |
APC | |
ZAP70 | |
70 kDa zeta-associated protein; 70 kDa zeta-chain associated protein; ADMIO2; EC 2.7.10.2; FLJ17670; FLJ17679; I79_004087; IMD48; kinase ZAP70; mrtle; mur; Selective T cell defect; Srk; STCD; STD; syk-related protein tyrosine kinase; syk-related tyrosine kinase; Tyrosine-protein kinase ZAP-70; Tyrosine-protein kinase ZAP-70;TZK; TZK; ZA70; Zap70; ZAP-70; Zeta chain associated protein kinase 70kDa; zeta chain of T cell receptor associated protein kinase; zeta chain of T cell receptor associated protein kinase 70; zeta chain of T cell receptor associated protein kinase 70kDa; zeta-chain (TCR) associated protein kinase; zeta-chain (TCR) associated protein kinase 70; zeta-chain (TCR) associated protein kinase 70kDa; zeta-chain associated protein kinase 70kDa; zeta-chain associated protein kinase, 70kD | |
Rabbit | |
Protein A/G | |
RUO | |
22637, 7535 | |
4° C | |
Liquid |
Flow Cytometry | |
Zap70Y319-A3 | |
PBS with 0.20% BSA and 0.09% sodium azide | |
P43403, P43404 | |
ZAP70 | |
A synthetic phospho-peptide corresponding to residues surrounding Tyr319/Tyr352 of human phospho Zap70/Syk. | |
100 Tests | |
Primary | |
Human, Mouse | |
Antibody | |
IgG κ |