Learn More
Cole-Parmer™ FF-200D-I Economical Flame Photometer
Product Code. 11485059
Change view
Click to view available options
Type:
Photometer Flame of Industry
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
11485059 Photometer Flame of Industry Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11485059 Supplier Cole-Parmer™ Supplier No. 8305505
  • ONLINE EXCLUSIVE PRICE
    5490.00€
    4294.09€ / pezzi 1
    Risparmia 1195.91€
    21% Sconto
View all promotions
Spedizione stimata: 15-12-2026
The FF-200D-I Economical Flame Photometer with automatic gas shutoff feature ensures operator safety.
  • Unit comes with Na+, K+, Li+, Ca+, and Ba+ filters.
  • Low temperature and single channel.
  • Use this economical flame photometer for routine determinations of sodium and potassium in industrial and educational applications.
  • The fine and coarse adjustment controls provide accurate measurements every time.
  • For safety, an automatic gas shutoff mechanism activates if the flame becomes accidentally extinguished.
Specifications
Voltage 230 VAC
Frequency 50/60 Hz
Type Photometer Flame of Industry