missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Phthalimide, 99%

Marca:  Alfa Aesar™ A12584.0C

Dettagli aggiuntivi : CAS : 85-41-6 Peso : 10.00000kg

Quantità 10000g

 vedi altre versioni di questo prodotto

Codice prodotto. 11404304

  • 258.00€ / 10000g


In stock, pronto per la spedizione: 5
Aggiungi al carrello

Descrizione

Descrizione

Applications
Phthalimide is a reagent used to transform allyl- and alkyl halides into protected primary amines. . It is used as a precursor to other organic compounds as a masked source of ammonia.

Solubility
Slightly soluble in water. Soluble in aqueous alkali hydroxides

Notes
Store at room temperature. Ensure good ventilation.
Specifica

Identificatori chimici

85-41-6
147.133
XKJCHHZQLQNZHY-UHFFFAOYSA-N
6809
isoindole-1,3-dione
C8H5NO2
MFCD00005881
phthalimide, 1h-isoindole-1,3 2h-dione, o-phthalimide, isoindoline-1,3-dione, o-phthalic imide, 1,3-isoindolinedione, benzoimide, 2-diazoindan-1,3-dione, phenylimide, phthalimid
CHEBI:38817
C1=CC=C2C(=C1)C(=O)NC2=O

Specifica

Phthalimide
165°C (329°F)
366°C
10000g
85-41-6
MFCD00005881
14,7373
Slightly soluble in water; Soluble in aqueous alkali hydroxides
C1=CC=C2C(=C1)C(=O)NC2=O
147.133
CHEBI:38817
99%
1.21
Odorless
232°C to 236°C
3.8
C8H5NO2
118522
phthalimide, 1h-isoindole-1,3 2h-dione, o-phthalimide, isoindoline-1,3-dione, o-phthalic imide, 1,3-isoindolinedione, benzoimide, 2-diazoindan-1,3-dione, phenylimide, phthalimid
XKJCHHZQLQNZHY-UHFFFAOYSA-N
isoindole-1,3-dione
6809
147.14
Videos
SDS Documenti