Learn More
Physostigmine Salicylate, TRC
Specifica
Specifica
| Materiale o nome chimico | Eserine Salicylate |
| CAS | 57-64-7 |
| Formula molecolare | C15 H21 N3 O2 . C7 H6 O3 |
| Quantità | 25 mg |
| Sinonimo | Physostigmine salicylate, (3aS,8aR)-1,2,3,3a,8,8a-Hexahydro-1,3a,8-trimethylpyrrolo[2,3-b]indol-5-yl methylcarbamate salicylate, Benzoic acid','2-hydroxy-','compd. with (3aS,8aR)-1,2,3,3a,8,8a-hexahydro-1,3a,8-trimethylpyrrolo[2,3-b]indol-5-yl N-methylcarbamate (1:1) (ACI), Benzoic acid','2-hydroxy-','compd. with (3aS,8aR)-1,2,3,3a,8,8a-hexahydro-1,3a,8-trimethylpyrrolo[2,3-b]indol-5-yl methylcarbamate (1:1) (9CI), Benzoic acid','2-hydroxy-','compd. with (3aS-cis)-1,2,3,3a,8,8a-hexahydro-1,3a,8-trimethylpyrrolo[2,3-b]indol-5-yl methylcarbamate (1:1) (ZCI), Physostigmine salicylate (6CI), Physostigmine','monosalicylate (8CI), Pyrrolo[2,3-b]indol-5-ol','1,2,3,3a,8,8a-hexahydro-1,3a,8-trimethyl-','methylcarbamate (ester)','(3aS,8aR)-','mono(2-hydroxybenzoate) (salt) (9CI), Pyrrolo[2,3-b]indol-5-ol','1,2,3,3a,8,8a-hexahydro-1,3a,8-trimethyl-','methylcarbamate (ester)','(3aS-cis)-','mono(2-hydroxybenzoate) (salt) (ZCI), Salicylic acid','compd. with physostigmine (1:1) (8CI), (-)-Physostigmine salicylate, Eserine salicylate, Isopto Eserine |
| InChI Formula | InChI=1S/C15H21N3O2.C7H6O3/c1-15-7-8-17(3)13(15)18(4)12-6-5-10(9-11(12)15)20-14(19)16-2;8-6-4-2-1-3-5(6)7(9)10/h5-6,9,13H,7-8H2,1-4H3,(H,16,19);1-4,8H,(H,9,10)/t13-,15+;/m1./s1 |
| SMILES | CNC(=O)Oc1ccc2N(C)[C@H]3N(C)CC[C@@]3(C)c2c1.OC(=O)c4ccccc4O |
| IUPAC Name | [(3aR,8bS)-3,4,8b-trimethyl-2,3a-dihydro-1H-pyrrolo[2,3-b]indol-7-yl] N-methylcarbamate;2-hydroxybenzoic acid |
| Molecular Weight (g/mol) | 413.47 |
| Peso formulazione | 413.1951 |