Learn More
MP Biomedicals™ Phytohemagglutinin, Phaseolus Vulgaris
Product Code. 11464196 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11464196 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11464196 Supplier MP Biomedicals™ Supplier No. 08791501
  • ONLINE EXCLUSIVE PRICE
    878.00€
    835.80€ / 25mg
    Risparmia 42.20€
    4% Sconto
 Restricted Item
Spedizione stimata: 22-12-2026
Phytohemagglutinin, Phaseolus Vulgaris is crude Phaseolus vulgaris-P lectin (PHA-P) from the Red Kidney Bean.
  • Crude Phaseolus vulgaris-P lectin (PHA-P) from the Red Kidney Bean.
  • Phytohemagglutinin PHA agglutinates the erythrocytes of all human blood groups as well as those of rabbit, dog, cat, mouse, sheep, and guinea pig.
  • The optimal amount of PHA required for the agglutination of erythrocytes varies with the blood from different individuals.
  • Protein or Enzyme Type: Other Proteins, Enzymes Peptides
Specifications
Quantity 25 mg
Form Lyophilized
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.