Learn More
Piceatannol, MedChemExpress
Product Code. 16443366
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16443366 25 mg 25mg
16431887 50 mg 50mg
16416216 100 mg 100mg
16405728 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16443366 Supplier Medchem Express Supplier No. HY13518
 Restricted Item
Spedizione stimata: 30-11-2026
MedChemExpress Piceatannol is a well-known Syk inhibitor and reduces the expression of iNOS induced by TNF.
Piceatannol is an effective agent for research of acute lung injury (ALI). Piceatannol is a naturally occurring polyphenolic stilbene found in various fruits and vegetables and exhibits anticancer and anti-inflammatory properties. Piceatannol induces apoptosis in DLBCL cell lines. Piceatannol induces autophagy and apoptosis in MOLT-4 human leukemia cells.
Specifications
Chemical Name or Material Piceatannol
CAS 10083-24-6
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C14H12O4
Quantity 25 mg
Synonym Astringenin trans-Piceatannol
Solubility Information DMSO : ≥ 31 mg/mL (126.92 mM) ∣H2O : 1 mg/mL (4.09 mM; Need ultrasonic)
SMILES OC1=CC=C(/C=C/C2=CC(O)=CC(O)=C2)C=C1O
Molecular Weight (g/mol) 244.24
Formula Weight 244.24
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only