missing translation for 'onlineSavingsMsg'
Learn More

(+)-Picfeltarraenin IA, TRC

Codice prodotto. 30410865
Cambia vista
Click to view available options
Quantità:
2.5 mg
missing translation for 'unitSize'
2.50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità unitSize
30410865 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30410865 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCP4363502.5MG

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Materiale o nome chimico Piefeltarraenin IA
CAS 97230-47-2
Formula molecolare C41 H62 O13
Quantità 2.5 mg
Sinonimo (3α,9β,10α,16α)-3-((2-O-(6-Deoxy-α-L-mannopyranosyl)-β-D-xylopyranosyl)oxy)-20,24-epoxy-16-hydroxy-9-methyl-19-norlanosta-5,23-diene-11,22-dione
InChI Formula InChI=1S/C41H62O13/c1-18(2)24-14-26(44)41(9,54-24)34-22(42)15-38(6)25-12-10-20-21(40(25,8)27(45)16-39(34,38)7)11-13-28(37(20,4)5)52-36-33(30(47)23(43)17-50-36)53-35-32(49)31(48)29(46)19(3)51-35/h10,14,18-19,21-23,25,28-36,42-43,46-49H,11-13,15-17H2,1-9H3/t19-,21?,22-,23-,25?,28-,29-,30+,31+,32+,33-,34?,35-,36+,38+,39-,40+,41+/m1/s1
SMILES CC(C)C1=CC(=O)[C@](C)(O1)C2[C@H](O)C[C@@]3(C)C4CC=C5C(CC[C@@H](O[C@@H]6OC[C@@H](O)[C@H](O)[C@H]6O[C@H]7O[C@H](C)[C@@H](O)[C@H](O)[C@@H]7O)C5(C)C)[C@]4(C)C(=O)C[C@]23C
IUPAC Name (2R)-2-[(3R,9R,13R,14S,16R)-3-[(2S,3R,4S,5R)-4,5-dihydroxy-3-[(2R,3S,4S,5S,6R)-3,4,5-trihydroxy-6-methyl-tetrahydropyran-2-yl]oxy-tetrahydropyran-2-yl]oxy-16-hydroxy-4,4,9,13,14-pentamethyl-11-oxo-1,2,3,7,8,10,12,15,16,17-decahydrocyclopenta[a]phenanthren-17-yl]-5-isopropyl-2-methyl-furan-3-one
Molecular Weight (g/mol) 762.92
Peso formulazione 762.419
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.