Learn More
Thermo Scientific™ Pierce™ 1-Step Transfer Buffer
Product Code. 15225346 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
200 mL
1 L
5 x 1 L
Unit Size:
1 L
200mL
5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15225346 5 x 1 L 5 L
15175204 1 L 1 L
15185204 200 mL 200mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15225346 Supplier Thermo Scientific™ Supplier No. 84731X5
 Restricted Item
In stock
Add to basket
Edge
Add to basket
Perform rapid semi-dry transfer of 10-300kDa proteins from polyacrylamide gels (SDS-PAGE) to nitrocellulose or PVDF membranes using the Pierce G2 Fast Blotter.
The Thermo Scientific™ Pierce 1-Step Transfer Buffer is designed for rapid semi-dry transfer of 10-300kDa proteins from polyacrylamide gels (SDS-PAGE) to nitrocellulose or PVDF membranes using the Pierce G2 Fast Blotter.
When used with the Pierce G2 Fast Blotter, this buffer provides effective gel-to-membrane transfer of proteins in 5 to 10 minutes with efficiency that is equal to, or better than, conventional Western blotting techniques. Pierce 1-Step Transfer Buffer is also compatible with other protein semi-dry transfer devices that are paired with a suitable high-current power supply. Such devices provide constant high current (1.3 to 5.0 amps) to rapidly transfer proteins via the high ionic strength conditions supplied by the transfer buffer.
Highlights:
  • Fast – high ionic strength formulation provides  5- to 10-minute protein transfer when used with compatible fast semi-dry blotting systems
  • Ready to use – supplied as a 1X solution that is stable and ready to use at room temperature
  • Optimized – designed for seamless and reliable performance with the Pierce G2 Fast Blotter
  • Compatible – effective with other semi-dry transfer devices equipped with a suitable high-current power supply
  • Economical – excellent performance without the special or costly consumables that are required by other fast semi-dry transfer devices
Required: Pierce G2 Fast Blotter (Part No. 62288) or other semi-dry transfer device that is equipped or paired with a capable high-current power supply.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Transfer Buffer
Buffer Pierce™ 1-Step Transfer buffer
Concentration 1 x
Description Pierce™ 1-Step Transfer buffer, 5 x 1L, Clear, Colorless, Liquid, 1X Concentration, 7 to 9 pH, SDS-PAGE Gel Type, Designed for Rapid Semi-Dry Transfer of 10 to 300kDa Proteins from Polyacrylamide Gels (SDS-PAGE) to Nitrocellulose or PVDF Membranes
Format Liquid
No. of Reactions 100 mini blots at 50mL each
Quantity 5 x 1 L
For Use With (Application) Protein gel electrophoresis