Learn More
Thermo Scientific™ DTT (Ditiotreitolo) Pierce™
Reduce disulfides in biological buffers and sample loading buffers with pure, crystalline dithiothreitol (DTT), the most popular protein disulfide reducing agent.
dietro preventivo
Descrizione
Thermo Scientific™ Pierce DTT (Dithiothreitol) is offered as 7.7mg × 48-tube microplates for convenient preparation of 100μL of 0.5M stocks for addition to protein samples for effective reduction of disulfide bonds.
Highlights:
- A water-soluble reagent that reduces disulfide bonds
- Maintains mono-thiols completely in the reduced state and reduces disulfide bonds quantitatively
- Specific and sensitive assay for disulfides using DTT with Ellman's Reagent