Learn More
Thermo Scientific™ Glycoprotein Standards Set for Pierce™ Glycoprotein Carbohydrate Estimation Kit
Product Code. 10760985 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 Protein
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10760985 5 Protein 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10760985 Supplier Thermo Scientific™ Supplier No. 23259
  • ONLINE EXCLUSIVE PRICE
    392.00€
    309.68€ / 1 sets
    Risparmia 82.32€
    21% Sconto
 Restricted Item
Spedizione stimata: 27-11-2026
Measure protein glycosylation as the percent of purified protein mass compared to included glycoprotein standards; uses periodic acid Schiff-base reagents (PAS).
Thermo Scientific™ Pierce Glycoprotein Carbohydrate Estimation Kit enables the amount of protein glycosylation to be measured as the percent of total purified protein mass.
The Glycoprotein Carbohydrate Estimation Kit includes six purified glycoprotein standards and the required assay reagents to effectively estimate the amount of oxidizable glycosylation (% carbohydrate by weight) of purified protein samples. Sugar groups in the glycoprotein sample are first oxidized with sodium meta-periodate to produce detectable aldehyde groups. Then the sample is reacted with the colorimetric Glycoprotein Detection Reagent. Absorbance at 550nm of the resulting purple reaction product is measured with a spectrophotometer or plate reader. Finally, the carbohydrate content in the glycoprotein is calculated by comparison to results from the five glycoprotein standards that are included in the kit.
Highlights:
  • Qualitative – easily identifies purified proteins as glycoproteins or samples as contaminated with sugars
  • Semi-quantitative – estimates the percent carbohydrate content (w/w) of purified glycoprotein by comparison to the included set of glycoprotein standards
  • Simple procedure – completed in less than 75 minutes; instructions include microplate and test tube protocols
  • Adaptable – standard curve format allows for the design of alternative tests for aldehydes and carbohydrate components
Includes:
Sodium periodate, assay buffer, detection reagent and a 5-vial set of glycoprotein standards
Recommended for:
Determine if a purified polyclonal antibody or other protein is glycosylated before attempting to perform carbohydrate-based conjugation or immobilization chemistries with hydrazide reagents; Characterize and classify purified protein fractions from an affinity procedure; Assess sugar and carbohydrate contamination in non-glycosylated protein samples
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Sufficient For: Preparing standard curves for glycoprotein carbohydrate estimation
• Lysozyme, lyophilized, 2.5 mg
• BSA, lyophilized, 2.5 mg
• Ovalbumin, lyophilized, 2.5 mg
• Apo-tranferrin, lyophilized, 2.5 mg
• Fetuin, lyophilized, 0.25 mg
• alpha-Acid Glycoprotein, 0.25 mg
Description Pierce Glycoprotein Standards Set
Assay Glycoprotein Standards
Detection Method Colorimetric
Product Line Pierce
Product Type Glycoprotein Carbohydrate Estimation
Specificity Not Target-Specific
Sufficient For 60 Tube Assays or 250 Microplate Assays
For Use With (Application) Internal Standard
For Use With (Equipment) Spectrophotometer, Microplate Reader
Quantity 5 Protein