Learn More
Thermo Scientific™ Pierce™ NeutrAvidin™ Coated Plates
Product Code. 10402905 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Black
Clear
White
No. of Wells:
96
384
Format:
Standard
Strip Plate
Plate Blocking:
Blocker™ BSA Buffer
SuperBlock™ Buffer
Unit Size:
Pack of 5
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color No. of Wells Format Plate Blocking Unit Size
10402905 White 384 Standard SuperBlock™ Buffer Pack of 5
10668534 Black 96 Standard Blocker™ BSA Buffer Pack of 5
15621995 Black 96 Standard SuperBlock™ Buffer Pack of 5
10093593 Black 96 Standard SuperBlock™ Buffer Pack of 5
10247673 Black 384 Standard SuperBlock™ Buffer Pack of 5
11597920 Clear 96 Strip Plate Blocker™ BSA Buffer Pack of 5
10732984 Clear 96 Strip Plate SuperBlock™ Buffer Pack of 5
10697764 Clear 96 Standard Blocker™ BSA Buffer Pack of 5
10165473 Clear 96 Standard SuperBlock™ Buffer Pack of 5
10165713 Clear 384 Standard SuperBlock™ Buffer Pack of 5
10195663 White 96 Standard Blocker™ BSA Buffer Pack of 5
10175423 White 96 Standard SuperBlock™ Buffer Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 10402905 Supplier Thermo Scientific™ Supplier No. 15401
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Spedizione stimata: 26-11-2026
Add to basket
Edge
Add to basket
Thermo Scientific™ Pierce NeutrAvidin Coated Plates are pre-blocked and ready-to-use polystyrene 96-well and 384-well plates for binding biotinylated antibodies or probes for ELISA and other target-specific assays.
Biotin-binding plates are ideal for applications when direct adsorption to polystyrene denatures antibodies or the target molecule. Thermo Scientific NeutrAvidin Protein is avidin that has been deglycosylated to prevent nonspecific lectin-binding. With a near-neutral pI (6.3), this results in lower nonspecific interactions in certain applications compared with streptavidin. NeutrAvidin Protein lacks the bacterial RYD sequence found on streptavidin, which eliminates nonspecific binding due to the RGD binding domain of adhesion receptors present in a variety of cells.
Thermo Scientific NeutrAvidin Protein Coated Plates are pre-blocked and ready-to-use polystyrene plates for binding biotinylated antibodies or probes for ELISA and other target-specific assays.
Features of NeutrAvidin Protein Coated Plates include:
  • White plates for chemiluminescent detection methods
  • Pre-blocked with Blocker BSA Buffer or SuperBlock Blocking Buffer
  • Easy and gentle immobilization of biotin-containing conjugates
  • Low nonspecific binding
  • No denaturing of the protein component of a conjugate upon binding
  • Ideal for binding small biotinylated hydrophilic molecules (e.g., peptides) that typically exhibit poor binding to polystyrene
  • Less expensive than streptavidin coated plates
  • Lowest nonspecific binding properties of all biotin-binding proteins
TRUSTED_SUSTAINABILITY
Specifications
Binding Property Standard Capacity, Affinity
Format Standard
Content And Storage Upon receipt store plates at 4°C in unopened pouches. Once opened, place unused plates in a resealable bag with desiccant and store at 4°C. Plates are shipped at ambient temperature.
Detection Method Chemiluminescence
Product Line NeutrAvidin Pierce
Color White
Product Type Microplate
Material Polystyrene
No. of Wells 384
Surface Treatment NeutrAvidin Protein
No. per Pack 5 Plates
Plate Blocking SuperBlock™ Buffer
Quantity 5 plates