Learn More
Thermo Scientific™ Pierce™ Recombinant Protein A/G, Alkaline Phosphatase Conjugated
Product Code. 11809240 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11809240 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11809240 Supplier Thermo Scientific™ Supplier No. 32391
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
Spedizione stimata: 04-12-2026
Detect antibodies, especially IgG isotypes, from many different species hosts with this purified, alkaline phosphatase- or peroxidase-conjugated recombinant Protein A/G.
Purified and conjugated forms of Thermo Scientific™ Pierce Recombinant Protein A/G enable probing and detection of antibodies, especially IgG isotypes, from many different species hosts in Western blotting, ELISA, IHC and other immunoassay protocols.
The extended Fc-binding properties of Protein A/G make it a popular tool in the investigation and purification of immunoglobulins. Protein A/G binds to all human IgG subclasses, IgA, IgE, IgM and to a lesser extent IgD; however, it does not bind mouse IgA, IgM or murine serum albumin. Protein A/G is an excellent tool for purification and detection of mouse monoclonal antibodies from IgG subclasses without interference from these other serum proteins. Individual subclasses of mouse monoclonals are most likely to have stronger affinity to this chimeric protein than to either Protein A or Protein G.
Highlights:
  • Contains four Fc-binding domains from Protein A and two from Protein G
  • Provides binding for all antibody species and subclasses recognized by either Protein A or Protein G
TRUSTED_SUSTAINABILITY
Specifications
Product Type Protein A, Alkaline Phosphatase Conjugate
Conjugate Alkaline Phosphatase
Quantity 0.5 mg
Product Line Pierce
Form Powder
Protein Subtype Protein A
Target Antibodies
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, but do not store above -20°C.