Learn More
Pilocarpine Hydrochloride, LoGiCal
Product Code. 30461492
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30461492 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30461492 Supplier LoGiCal Supplier No. LGCFOR0338.00
 Restricted Item
Spedizione stimata: 30-11-2026
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Pilocarpine Hydrochloride
CAS 54-71-7
Analyte or Component Names Pilocarpine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C11 H16 N2 O2 . Cl H
Certifications/Compliance ISO 9001
Synonym Pilocarpine Hydrochloride, (3S,4R)-3-Ethyldihydro-4-[(1-methyl-1H-imidazol-5-yl)methyl]-2(3H)-furanone monohydrochloride, (3S-cis)-3-Ethyldihydro-4-[(1-methyl-1H-imidazol-5-yl)methyl]-2(3H)-furanone monohydrochloride, Pilocarpine monohydrochloride, (+)-Pilocarpine hydrochloride, Adsorbocarpine, Almocarpine, Isopto Carpine, NSC 5746, Pilocar SMP, Pilocarpal, Pilocarpine chloride, Pilocarpine hydrochloride, Pilocarpine muriate, Pilocarpinium chloride, Pilocel, Pilovisc, Salagen, Vistacarpin N
InChI Formula InChI=1S/C11H16N2O2.ClH/c1-3-10-8(6-15-11(10)14)4-9-5-12-7-13(9)2;/h5,7-8,10H,3-4,6H2,1-2H3;1H/t8-,10-;/m0./s1
Solution Type Neat
SMILES Cl.CC[C@H]1[C@@H](Cc2cncn2C)COC1=O
IUPAC Name (3S,4R)-3-ethyl-4-[(3-methylimidazol-4-yl)methyl]oxolan-2-one;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 244.72
Quantity 10 mg
Formula Weight 244.0979 g/mol