missing translation for 'onlineSavingsMsg'
Learn More

PIPES, 0.5M buffer soln., pH 6.0, Thermo Scientific™

Catalog Number p-7042346
Click to view available options
Quantità:
100 mL
250 mL
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
5625-37-6
C8H18N2O6S2
302.36
MFCD00006159
IHPYMWDTONKSCO-UHFFFAOYSA-N
79723
CHEBI:44933
2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid
C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
5625-37-6
C8H18N2O6S2
302.36
MFCD00006159
IHPYMWDTONKSCO-UHFFFAOYSA-N
79723
CHEBI:44933
2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid
C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico PIPES buffer soln.
Colore Colorless
Concentrazione 0.5 M
pH 6.0
Forma fisica Liquid
Quantità 100 mL
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'