Learn More
Sartorius™ Pipette Calibration Set
Product Code. 14880701 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
ME and CPA Analytical Balances
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
14880701 ME and CPA Analytical Balances Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 14880701 Supplier Sartorius™ Supplier No. YCP04HW
 Restricted Item
In stock
Used for ME and CPA analytical balances.
  • Used for ME and CPA analytical balances
  • Consists of moisture trap and all needed adapters
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) ME and CPA Analytical Balances