Learn More
Hamilton™ Pipettor tip, 300μL rocket CO-RE non-sterile, black, for Microlab STAR automated pipetting system
Codice prodotto. 15127416
Cambia vista
Click to view available options
Volume (sistema metrico):
300 μL
Dimensione della confezione:
pezzi 4800
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Volume (sistema metrico) unitSize
15127416 300 μL pezzi 4800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 15127416 Fornitore Hamilton™ N. del fornitore 235974
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
The Hamilton consumables range is produced in an ISO Class 8 Clean Room, based on ISO 14644 standards as well as ISO 9001 and ISO 13485. Hamilton goes beyond relying on just statistical analysis of quality to assure the highest product quality, e.g. each CO-RE tip is checked for concentricity of its orifice and the absence of flash from the moulding process. TüV Rheinland certifies that our installed Quality Management System has been assessed and found to be in accordance with the requirements of the high performance standards for design and manufacturing of disposable products. Most consumables meet the EC directives CE and IVD. To meet the highly sophisticated needs of laboratories in the Genomics, Proteomics, Cellomics, Forensic and Drug Discovery fields, Hamilton has considerably refined the production requirements for sterile/non-sterile disposable products. The superior biological purity of CO-RE pipettor tips, tubes, and reagent containers is guaranteed by a controlled production process. Skilled personnel, along with automated clean room production, enable contamination-free manufacturing. At regular intervals the production batch is subjected to a highly sensitive PCR method to secure compliance with the defined purity criteria of Dnase/Rnase, pyrogen and PCR inhibitor-free. Hamilton has introduced a new purity grade especially for forensic work, to eliminate the residual risk of contamination of clean products. Along with the strictest purity requirements during the production process, an additional step is performed during the manufacturing and packing of disposable end products. This treatment ensures the effective removal of contaminants by physically destroying any DNA molecules. In accordance with Forensic Science International (FSI), Hamilton has validated the treatment process to prove the effectiveness of eliminating amplifiable DNA. Considering that just one contamination event from extraneous DNA during forensic investigation may result in incorrect DNA profiling, rendering evidence worthless, Hamilton's commitment to purity is of the utmost importance.
  • Automated clean room tip production and quality control as part of the Hamilton consumables range
  • Hamilton biological-purity tested products include Dnase/Rnase, pyrogen (endotoxin) and PCR inhibitor-free materials
  • CE IVD certified where applicable
  • The Hamilton range of filter pipette tips prevent sample carry-over and aerosol contamination
  • Economical, standardised tip packaging include tip trays in SBS format and a nested tip rack reload system
  • Full traceability and easy tip volume identification with a colour-coded barcode label, where applicable
  • Sterility of products validated according to ISO 11137
Specifica
Volume (sistema metrico) 300 μL
Sterilità Non sterile
Colore Black
Da utilizzare con (apparecchiatura) Microlab STAR Automated Pipetting System