missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKC gamma Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2F4B9, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66429-1-IG-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role in cells.Specifica
PKC gamma | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P05129, P63318, P63319 | |
Prkcg | |
PKC gamma Fusion Protein Ag5910 | |
20 μL | |
Primary | |
Porcine, Mouse, Human, Rat | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot | |
2F4B9 | |
Unconjugated | |
PRKCG | |
PKC gamma, PKCC, PKCG, PRKCG, Protein kinase C gamma type, protein kinase C, gamma, SCA14 | |
Mouse | |
Protein A | |
RUO | |
100518074, 18752, 24681, 5582 | |
-20°C | |
Liquid |