Learn More
Brannan™ Plant Light and Moisture Meter
Product Code. 13120443
Change view
Click to view available options
Includes:
A plant reference list
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
13120443 A plant reference list Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13120443 Supplier Brannan™ Supplier No. 13/715/0
  • ONLINE EXCLUSIVE PRICE
    25.60€
    22.50€ / pezzi 1
    Risparmia 3.10€
    12% Sconto
Spedizione stimata: 10-12-2026
Tests both for moisture content in soil or compost and for light falling onto a plant.
  • Supplied completer with guide to light and watering requirements of plant
Specifications
Type Plant Light and Moisture Meter
Height (Metric) 10 cm
Includes A plant reference list
Weight (Metric) 0.093 kg
Width (Metric) 6.8 cm