missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLAP Mouse anti-Human, Clone: 2B7C9, Proteintech
Mouse Monoclonal Antibody
140.00€ - 399.00€
Specifica
Antigene | PLAP |
---|---|
Clone | 2B7C9 |
Concentrazione | 0.6 mg/mL |
Applicazioni | Immunohistochemistry (Paraffin), Western Blot |
Classificazione | Monoclonal |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
16807375
|
Proteintech
60294-1-IG-20UL |
20 μL |
140.00€
20µL |
Spedizione stimata: 08-05-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
16897365
|
Proteintech
60294-1-IG-150UL |
150 μL |
399.00€
150µL |
Spedizione stimata: 08-05-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
Descrizione
Placental Alkaline Phosphatase plays an important role in the regulation of specific inflammatory disease processes. There are at least four distinct but related alkaline phosphatases: intestinal, placental, placental-like, and liver/bone/kidney for this form of alkaline phosphatase have been well characterized.Placental Alkaline Phosphatase reacts with a membrane-bound isoenzyme (Regan and Nagao type) of Placental Alkaline Phosphatase (PLAP) occurring in the placenta during the 3rd trimester of gestation. Placental Alkaline Phosphatase is useful in the identification of testicular germ cell tumors. Unlike germ cell tumors, PLAP-positive somatic cell tumors uniformly express epithelial membrane antigen (EMA). A proposed function of Placental Alkaline Phosphatase is matrix mineralization; however, mice that lack a functional form of this enzyme show normal skeletal development. Placental Alkaline Phosphatase has been linked directly to hypophosphatasia, a disorder that is characterized by hypercalcemia and includes skeletal defects. The character of hypophosphatasia can vary, however, depending on the specific mutation since this determines age of onset and severity of symptoms.Specifica
PLAP | |
0.6 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ALP, ALPP, PALP, Placental alkaline phosphatase, PLAP, PLAP 1 | |
ALPP | |
IgG1 | |
Protein G | |
Antibody |
2B7C9 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P05187 | |
250 | |
PLAP Fusion Protein Ag13361 | |
Primary | |
-20°C | |
ALPP |