Learn More
Macherey-Nagel™ Neutralization Buffer NEU
Product Code. 11902182
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11902182 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11902182 Supplier Macherey-Nagel™ Supplier No. 740348.1000
  • ONLINE EXCLUSIVE PRICE
    105.00€
    95.03€ / 1000mL
    Risparmia 9.97€
    9% Sconto
 Restricted Item
In stock
Neutralization Buffer NEU to be used with NucleoBond Xtra Midi/Maxi kits.
Specifications
Chemical Name or Material Neutralization Buffer
For Use With (Application) For Plasmid DNA Purification
Quantity 1000 mL