Learn More
3M™ Plastic Sweatband
Product Code. 17267206 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
3M Headgear Combinations G500 Series
All 3M™ Hard Hats
Unit Size:
Pack of 10
Pack of 20
Pack of 250
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17267206 All 3M™ Hard Hats Pack of 250
15729867 3M Headgear Combinations G500 Series Pack of 10
15739957 All 3M™ Hard Hats Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17267206 Supplier 3M™ Supplier No. 7000108029
  • ONLINE EXCLUSIVE PRICE
    1040.00€
    826.80€ / pezzi 250
    Risparmia 213.20€
    20% Sconto
Spedizione stimata: 05-02-2027
3M™ Plastic Sweatband suitable for use with 3M™ hard hats and headgear combinations.
  • Replacement plastic sweatband suitable for use with 3M™ Hard Hats and 3M™ G500 Headgear Combinations
  • Quick and easy to install
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE Approved
Color Black
For Use With (Equipment) All 3M™ Hard Hats
Material Plastic
Product Type Sweatband