Learn More
Kartell™ Bottiglie graduate in polietilene con collo largo
Descrizione
- Permanent graduation printed in relief externally
- Supplied with under cap and screw cap
- Will withstand considerable pressure before rupture
- Secured by tagging points on bottle and cap
- Confidential notes can be placed in the space between insert sealing plug and cap
- Suitable for foodstuff according to EC Regulation 10/2011
- Easy grip caps can be wired down for security
- Accurate graduation and semi-transparent walls allow a clear and precise reading of the content
Specifica
Specifica
| Capacità (sistema metrico) | 100 mL |
| Colore | Opaque |
| Materiale | Polyethylene |
| Sterilità | Non sterile |
| Graduazioni | 20 mL |
| Closure Included | Sì |
| Graduated | Yes |
| Bocca | Larga |
| Altezza (sistema metrico) | 89 mm |
| Da utilizzare con (apparecchiatura) | Solids and Liquids |
| Vedi altri risultati |