Learn More
Invitrogen™ Collibri™ Library Amplification Master Mix
Codice prodotto. 15950523 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
N. di reazioni:
50
250
Dimensione della confezione:
50 reazioni
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. N. di reazioni Dimensione della confezione
15950523 50 50 reazioni
15960523 250 50 reazioni
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 15950523 Fornitore Invitrogen™ N. del fornitore A38539050
Focus on Regenerative Therapies
Usa il codice promozionale 28027
Visualizza tutte le promozioni
 Articolo soggetto a restrizioni
In stock
Invitrogen™ Platinum SuperFi Library Amplification Master Mix is a ready-to-use solution designed for amplification of next-generation sequencing (NGS) libraries.
Master mix includes Platinum SuperFi™ DNA Polymerase in combination with a proprietary reaction buffer containing all necessary components for efficient library amplification with minimal bias and superior accuracy.
 Features include:
  • High fidelity—minimal error rates similar to PCR-free methods
  • High efficiency—high yields from minute input amounts
  • Minimal bias—uniform amplification regardless of GC content
  • Platinum SuperFi™ DNA Polymerase has both 5’ to 3’ polymerase and 3’ to 5’ exonuclease (proofreading) activities, but lacks 5’ to 3’ exonuclease activity. Exceptionally strong proofreading activity ensures amplification of NGS libraries with supreme sequence accuracy, producing NGS error rates similar to PCR-free methods
  • Proprietary reaction buffer contains all necessary PCR components and additives to enable efficient and uniform library amplification regardless of GC content, helping improve coverage across GC- and AT-rich sequences and other complex regions. High amplification efficiency enables the use of fewer cycles to achieve the desired yield
  • Platinum hot-start technology inhibits DNA polymerase activity at ambient temperatures, allowing room temperature reaction setup and storage of pre-assembled PCR reactions for up to 24 hours prior to the PCR
  • Platinum SuperFi™ Library Amplification Master Mix is supplemented with an inert blue dye that provides a visual aid when pipetting and decreases the risk of pipetting errors during reaction setup
  • For increased convenience, we offer Platinum SuperFi™ Library Amplification Master Mix with Primer Mix, where the master mix is supplied with a 10X Primer Mix that targets the P5 and P7 regions of Illumina adapters and that is supplemented with an inert yellow dye. Mixing both components in a PCR reaction turns the final PCR solution green, further decreasing the risk of pipetting errors
TRUSTED_SUSTAINABILITY
Specifica
Content And Storage 1 x 1.25 mL Collibri Library Amplification Master Mix (2X)
Store at -20°C.
Tipo di campione DNA, RNA
Fedeltà (rispetto a taq) ≥100
Prestazioni PCR arricchito GC High
Da utilizzare con (applicazione) Sequencing
Hot Start Built-In Hot Start
N. di reazioni 50
Sporgenza Blunt
Linea di prodotti Collibri
Product Type Library Amplification Master Mix
Quantità 50 reactions
Reaction Format SuperMix or Master Mix
Shipping Condition Dry Ice
Durata del test Standard
Forma Liquid