missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLD3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17327-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descripción
PLD3 is a ingle-pass type II membrane protein. It belongs to the phospholipase D family. PLD3 contains 2 PLD phosphodiesterase domains. The exact function of PLD3 remains unknown.Especificaciones
PLD3 | |
Polyclonal | |
Unconjugated | |
PLD3 | |
Choline phosphatase 3, HindIII K4L homolog, HU K4, Phospholipase D3, PLD 3, PLD3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18807, 23646, 361527 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O35405, Q5FVH2, Q8IV08 | |
Pld3 | |
PLD3 Fusion Protein Ag11306 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |