Learn More
BTX™ Pliers for High-Throughout Electroporation Microplate Handler
Product Code. 15791088
Change view
Click to view available options
For Use With (Equipment):
BTX High Throughput Electroporation Plate Handler
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15791088 BTX High Throughput Electroporation Plate Handler Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15791088 Supplier BTX™ Supplier No. 5012017
 Restricted Item
Spedizione stimata: 09-12-2026
Pliers for High-Throughout Electroporation Microplate Handler is an accessory for moving or replacing gold contact pins.
Features and Benefits:
  • Made with a specific plasticized tooth to grip the gold contact pins, but not indent or damage the integrity of the voltage conductivity, these pliers come as standard with High-Throughput plate handler, but can also be bought separately.
Specifications
For Use With (Equipment) BTX High Throughput Electroporation Plate Handler
Grade General Precision
Autoclavable Not Autoclavable