missing translation for 'onlineSavingsMsg'
Learn More

PLOD2 Mouse anti-Human, Clone: 1H9E1, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66342-1-IG-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898125

  • 140.00€ / 20µL
Spedizione stimata: 14-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

The protein encoded by this gene is a membrane-bound homodimeric enzyme that is localized to the cisternae of the rough endoplasmic reticulum. The enzyme (cofactors iron and ascorbate) catalyzes the hydroxylation of lysyl residues in collagen-like peptides. The resultant hydroxylysyl groups are attachment sites for carbohydrates in collagen and thus are critical for the stability of intermolecular crosslinks. Some patients with Ehlers-Danlos syndrome type VIB have deficiencies in lysyl hydroxylase activity. Mutations in the coding region of this gene are associated with Bruck syndrome. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifica

Specifica

PLOD2
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O00469
PLOD2
PLOD2 Fusion Protein Ag5779
20 μL
Primary
Human
Antibody
IgA
Western Blot, Immunohistochemistry (Paraffin)
1H9E1
Unconjugated
PLOD2
LH2, Lysyl hydroxylase 2, PLOD2, TLH
Mouse
Affinity Chromatography
RUO
5352
-20°C
Liquid
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali