Learn More
Motic™ Polarizing Set, 18/28 LED Series
Descrizione
- Improve image quality obtained with birefringent materials when compared to other techniques such as darkfield and brightfield illumination, differential interference contrast, phase contrast, Hoffman modulation contrast, and fluorescence.
- Useful for both quantitative and qualitative studies targeted at a wide range of anisotropic specimens.
- Polarized light microscopy is capable of providing information on absorption color and optical path boundaries between minerals of differing refractive indices, in a manner similar to brightfield illumination, but the technique can also distinguish between isotropic and anisotropic substances.
- Contrast-enhancing technique exploits the optical properties specific to anisotropy and reveals detailed information concerning the structure and composition of materials that are invaluable for identification and diagnostic purposes.
Specifica
Specifica
| Tipo | Polarizer, Analyzer |
| Descrizione | Permits 360° of rotation to accommodate observation demands. |
| Da utilizzare con (apparecchiatura) | 18/28 LED Series Microscopes |