Learn More
POLR1A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA556766
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: SNYEVIMKSQ HLHEVLSRAD PKKALHHFRA IKKWQSKHPN TLLRRGAFLS YSQKIQEAVK ALKLESENRN GRSPGTQEML RMWYELDEES RRKYQ Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 76%.
POLR1A Polyclonal antibody specifically detects POLR1A in Human samples. It is validated for ChIP Assay, Immunohistochemistry.Specifica
POLR1A | |
Polyclonal | |
Unconjugated | |
POLR1A | |
194kDa; 3010014K16Rik; A190; A194; AFDCIN; DNA-directed RNA polymerase I A; DNA-directed RNA polymerase I largest subunit; DNA-directed RNA polymerase I subunit A; DNA-directed RNA polymerase I subunit A1; DNA-directed RNA polymerase I subunit RPA1; mRPA1; Polr1a; polymerase (RNA) I polypeptide A; polymerase (RNA) I polypeptide A, 194kDa; polymerase (RNA) I subunit A; RNA polymerase 1-4; RNA polymerase 1-4 (194 kDa subunit); RNA polymerase I (194 kDa subunit); RNA polymerase I 194 kDa subunit; RNA polymerase I subunit A; RNA polymerase I subunit A1; Rpa1; RPA194; RPO14; RPO1-4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25885 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ChIP Assay, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95602 | |
POLR1A | |
Recombinant protein corresponding to Human POLR1A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only