Learn More
Poly(I:C) (~90%), TRC
Codice prodotto. 30410498
Cambia vista
Click to view available options
Quantità:
10 mg
250 mg
1 g
Dimensione della confezione:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30410498 10 mg 10mg
30410896 250 mg -
30423843 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Codice prodotto. 30410498 Fornitore Toronto Research Chemicals N. del fornitore TRCP68807510MG
 Articolo soggetto a restrizioni
Spedizione stimata: 13-08-2026
Aggiungi al carrello
Aggiungi al carrello
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 24939-03-5
Formula molecolare H2(C10 H11 N4 O8 P)n . H2(C9 H12 N3 O8 P)n
Molecular Weight (g/mol) 671.4
Sinonimo 5'-Inosinic Acid Homopolymer Complex with 5'-Cytidylic Acid Homopolymer, 5'-Inosinic Acid Polymers Complex with 5'-Cytidylic Acid Polymers, 5'-Cytidylic Acid Homopolymer Complex with 5'-Inosinic Acid Homopolymer, 5'-Cytidylic Acid Polymers Complex with 5'-Inosinic Acid Polymers, 5'-Inosinic Acid Homopolymer-5'-cytidylic Acid Homopolymer Complex, NSC 120949, NSC 123233, NSC 128576, Poly(C).poly(I), Poly(C:I), Poly(I)-poly(C), Poly(I).poly(C), Poly(I).poly(C) Complex, Poly(cytidylic acid)-poly(inosinic acid), Poly(rI)-poly(rC), Poly(rI).poly(rC), Poly(rI):poly(rC), Poly(rI:rC), Polycytidylic-polyinosinic Acid, Polyinosinate-polycytidylate Complex, Polyinosinate:polycytidylate, Polyinosine-polycytidylic Acid, Polyinosinic Acid:polycytidylic Acid, Polyinosinic Acid:polycytidylic Acid Complex, Polyinosinic-polycytidylic Acid, Polyinosinic-polycytidylic Acid Complex, Polyribocytidylic Acid-polyriboinosinic Acid Complex
SMILES O[C@@H]1[C@H](O)[C@@H](COP(O)(O)=O)O[C@H]1N2C(NC=NC3=O)=C3N=C2.O[C@@H]4[C@H](O)[C@@H](COP(O)(O)=O)O[C@H]4N5C=CC(N)=NC5=O
Specifica
Materiale o nome chimico Poly(I:C)
Quantità 10 mg
InChI Formula InChI=1S/C10H13N4O8P.C9H14N3O8P/c15-6-4(1-21-23(18,19)20)22-10(7(6)16)14-3-13-5-8(14)11-2-12-9(5)17;10-5-1-2-12(9(15)11-5)8-7(14)6(13)4(20-8)3-19-21(16,17)18/h2-4,6-7,10,15-16H,1H2,(H,11,12,17)(H2,18,19,20);1-2,4,6-8,13-14H,3H2,(H2,10,11,15)(H2,16,17,18)
Peso formulazione 671.099