Learn More
Scilabub™ Polyamide Fiber Protective Gloves
Product Code. 12302239
Change view
Click to view available options
Size:
Small
Medium
Large
Unit Size:
1 pair
Pack of 2
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12302239 Medium 1 pair
11883841 Medium Pack of 2
11873801 Small Pack of 2
12921185 Small 1 pair
11893841 Large 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12302239 Supplier Scilabub™ Supplier No. GAU/NM
  • ONLINE EXCLUSIVE PRICE
    244.00€
    209.75€ / 1 paia
    Risparmia 34.25€
    14% Sconto
Spedizione stimata: 21-12-2026
These Nomex™ polyamide heat resistant gauntlet gloves are resistant to 260°C (working temperature, dry heat only) with knitted fabric construction for extra comfort.
  • Heat-resistant (260°C working temperature, dry heat only)
  • Low thermal conductivity
  • Asbestos free
  • Double thickness
  • Knitted fabric for comfort
  • Does not shrink or melt
  • Unaffected by washing
  • Hard wearing
  • Conform to EN338: 2 X 4 X and EN407: 41324X
Specifications
Certifications/Compliance CE Mark
Color Red
Material Polyamide
Powdered Powder Free
Product Type Protective Gloves
Size Medium
Sterility Non-sterile
Resistant To Heat, Chemical
Length (Metric) 52 cm