Learn More
R&D Systems™ Polybrene
Product Code. 18717326
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18717326 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18717326 Supplier R&D Systems™ Supplier No. 7711/10
  • ONLINE EXCLUSIVE PRICE
    165.94€
    148.05€ / 10mg
    Risparmia 17.89€
    10% Sconto
 Restricted Item
In stock
Add to basket
Edge
Add to basket
A Tocris product, part of R&D Systems. Viral transduction enhancer
Polybrene is a cationic polymer that improves lentiviral and adenoviral transduction efficiency in mammalian cells in vitro. It also enhances DNA transfection in many types of cells. It has a neutralizing effect on negatively charged DNA and viral particles which facilitates their attachment and entry into host cells. Polybrene is suitable for use with CRISPR/Cas9 protocols.
Polybrene is a lyophilized solid and can be prepared as a stock solution by adding 1 mL of sterile ultra-pure water directly to the vial (final stock concentration 10 mg/mL).
For further information view the product protocol .
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,5-Dimethyl-1,5-diazaundecamethylene polymethobromide
CAS 28728-55-4
Quantity 10 mg
Product Line Tocris
Target Viral Transduction Enhancers
Purity 0.95