Learn More
Brady™ Polyester Glow-in-the-dark Right Footprint
Product Code. 11771875 Shop All Brady Products
Change view
Click to view available options
Color:
Photoluminescent
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11771875 Photoluminescent Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11771875 Supplier Brady™ Supplier No. 055359
  • ONLINE EXCLUSIVE PRICE
    10.10€
    9.06€ / pezzi 1
    Risparmia 1.04€
    10% Sconto
In stock
Use the Polyester Glow-in-the-dark Right Footprint together with the left foot to create a visual pathway to aid a fast exit during power outages and emergency situations.
  • Use together with the left foot to create a visual pathway to aid a fast exit during power outages and emergency situations.
Specifications
Alternate Part Number B324 VINYL FOOTPRINT-RIGHT 12 in. GLOW 1 PC
Certifications/Compliance ASTM E-2072-00 / ASTM E-2073-00, ASTM E-162, ASTM E-648, ASTM E-662, IMO Resolution A.752(18),, ISO/CD 15370, Marine Safety Committee MSC.27(61), DIN 67 510 (Parts 1 - 4) and PSPA Standard 002 Part 2. Meets OSHA 1910.37, Lloyd's Register SAS F050294.p
Description Glow-in-the-dark footprint - Right foot
For Use With (Application) Use Together With the Left Foot to Create a Visual Pathway
Height (English) 12 in.
Height (Metric) 304.8 mm
Packaging Quantity 1 Ea.
Temperature Range (English) -40°F to +176°F
Temperature Range (Metric) -40°C to +80°C
Thickness 0.2 mm
Width (English) 3.5 in.
Width (Metric) 88.9 mm
Adhesive Type Permanent Acrylic
Color Photoluminescent
Material Polyester