Learn More
Cole-Parmer™ Cella di polietilene dotata di guanti interni
Codice prodotto. 12368627 Sfoglia Tutto Cole-Parmer Prodotti
Cambia vista
Click to view available options
Lunghezza (sistema metrico):
431.8 mm
508 mm
685.8 mm
939.8 mm
Larghezza (sistema metrico):
431.8 mm
508 mm
635 mm
685.8 mm
939.8 mm
Altezza (sistema metrico):
279.4 mm
304.8 mm
381 mm
457.2 mm
939.8 mm
Dimensione della confezione:
pezzi 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Lunghezza (sistema metrico) Larghezza (sistema metrico) Altezza (sistema metrico) Dimensione della confezione
12368627 508 mm 508 mm 304.8 mm pezzi 6
12318627 431.8 mm 431.8 mm 279.4 mm pezzi 6
12328627 431.8 mm 685.8 mm 304.8 mm pezzi 6
12338627 685.8 mm 685.8 mm 381 mm pezzi 6
12358627 939.8 mm 635 mm 939.8 mm pezzi 6
12348627 685.8 mm 939.8 mm 457.2 mm pezzi 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Codice prodotto. 12368627 Fornitore Cole-Parmer™ N. del fornitore 0440840
  • ONLINE EXCLUSIVE PRICE
    439.00€
    353.18€ / pezzi 6
    Risparmia 85.82€
    19% Sconto
Visualizza tutte le promozioni
 Articolo soggetto a restrizioni
Spedizione stimata: 25-01-2027
Protect yourself and your materials quickly without giving up lab space with these glove chambers. Glove Bag™ Inflatable Glove Chambers establish high-purity working atmosphere in minutes. Inflatable chambers effectively eliminate oxygen and moisture during material manipulation operation.
An alternative to standard glove boxes—these durable, polyethylene glove chambers are reusable, yet priced to be disposable. Chambers are easy to use—just slide your equipment through the sleeves, seal, connect the gas port (accepts 3⁄8'' ID tubing) to your gas supply, and inflate.
  • Disposable chambers simplify cleanup
  • Two equipment sleeves and two rear-mounted gas ports for added versatility
  • Glove bags have a 2.5 mil thickness and are made of polyethylene
  • Glove bags are not autoclaveable, nor are they sterile
  • One pair of cotton gloves, tubing coupler and a closure included
This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Specifica
Materiale Polietilene
Altezza (sistema metrico) 304.8 mm
Include 1 Pair Of Cotton Gloves, Tubing Coupler, and a Closure
Lunghezza (sistema metrico) 508 mm
Quantità 6/Pk.
Larghezza (sistema metrico) 508 mm