missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene glycol methyl ether methacrylate, M.W. 13,000, Thermo Scientific Chemicals
Click to view available options
Quantità:
250 mg
1 g
5 g
Descrizione
Polyethylene glycol methyl ether methacrylate, is used as Emulsion polymers, cosmetics, construction, emulsifiers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Materiale o nome chimico | Polyethylene glycol methyl ether methacrylate |
Colore | White |
Nota nome | M.W. 13,000 |
Forma fisica | Granular Solid |
Informazioni di solubilità | Soluble in water. |
Quantità | 250 mg |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'