Learn More
Fisherbrand™ Polyethylene Push-in Caps for Glass Specimen Vials
Product Code. 15364789 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Closure Size:
8 mm
10 mm
22 mm
Unit Size:
Pack of 300
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Size Unit Size
15364789 8 mm Pack of 300
15354789 22 mm Pack of 300
15374789 10 mm Pack of 300
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 15364789
Spedizione stimata: 11-12-2026
Add to basket
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Fit securely into place to seal glass specimen vials. Fisherbrand™ Polyethylene Push-in Caps for Glass Specimen Vials are available in a variety of sizes specifically designed to work with Fisherbrand™ Push-in Cap Glass Specimen Vials.
  • Specifically designed for use with Fisherbrand™ Push-in Cap Glass Specimen Vials
  • Polyethylene construction, natural color
  • Available in a variety of sizes to match vial specifications
  • Push-in cap specimen vials are suitable for media, diagnostic, storage, display, and sample collection applications
Specifications
Color Natural
Material Polyethylene
Closure Type Push-in Cap
Closure Size 8 mm