Learn More
Masterflex™ Polyethylene Straight Connector, pack of 100
Designed for quick and easy assembly without tools
Marca:  Masterflex™ 30626-50
Dettagli aggiuntivi : Peso : 0.11000kg
Codice prodotto. 15333568
  • 117.00€ / pezzi 100
Spedizione stimata: 23-07-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Adapts from a NPT male thread to a hose barb
  • Barbed fittings are designed for quick and easy assembly without tools
  • Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30psi)
  • Available in packs of ten
Specifica
Specifica
Straight Connector
Not Autoclavable
5/32 in.
5/32 in.
Translucent
Barb Connection
HDPE
5/32 in.
Ethylene Oxide