Learn More
Hirschmann™ Polypropylene Thread Adapter
Descrizione
- 9313505/9313506: For use with ceramus™ -classic.
- 9313527: For use with ceramus™, solarus™ and opus™.
Specifica
Specifica
| Product Type | Adapter |
| Materiale | Polypropylene |
| Dimensioni filo | A 32 to A 38 |