Learn More
Brady™ Polypropylene Cleats For 120/277V Lockout
Product Code. 15623507 Shop All Brady Products
Change view
Click to view available options
Color:
Red
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15623507 Red Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15623507 Supplier Brady™ Supplier No. 065404
  • ONLINE EXCLUSIVE PRICE
    15.25€
    13.94€ / pezzi 6
    Risparmia 1.31€
    8% Sconto
In stock
Specifications
Product Type Cleats
Alternate Part Number LOCKOUT CLEAT 277 VOLT GENERAL USE PK6
Color Red
Description Extra Cleats for 120/277V Lockout
For Use With (Equipment) For 120/277V Lockout
Quantity 6 Pk.