Learn More
Chemsplash Polypropylene Lab/Visitors Coat White with Velcro Closure
Descrizione
Visitors Polypropylene Disposable Lab Coat
- Material: 30gsm Polypropylene Fabric
- Features:
- Velcro Closure
- With Fold Over Collar
- Elasticated Cuffs
- Ideal for Non-Hazardous Environments
- Ideal for Food Industry
- PFAS Free
The Visitors Polypropylene Disposable Lab Coat is designed with a velcro front closure and is suitable for visitors, site inspections, or general wear in minimal risk environments. It is ideal for use in the food industry or other non-hazardous settings. This product is suitable for use in BRC-approved work areas and the pharmaceutical industry.
This product does not contain: PFAS, RoHS, POP, TSCA, REACH, or any conflict materials.
Specifica
Specifica
| Product Type | Visitor Lab Coat |
| Size | Small |
| Sesso | Unisex |
| Materiale | Polypropylene |
| Colore | White |
| Tipo di chiusura | Hook and Loop |
| Stile polso | Elastic |
| Monouso | Single-use |
| Sterilità | Non-sterile |
| Resistenza | Particulates |
| Vedi altri risultati |