Learn More
Masterflex™ Raccordo Luer in polipropilene
Codice prodotto. 15305641
Cambia vista
Click to view available options
Da utilizzare con:
Female Luer x 1/4 to 28 in. UNF
Female Luer x Female Luer, Elbow
Female x Female x Female
Male LL to 1/4 to 28 UNF Thread
Male LL to 10 to 32 Thread
Male x Female x Female
Dimensione della confezione:
25 confezioni
pezzi 25
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Da utilizzare con unitSize
15305641 Female Luer x Female Luer, Elbow pezzi 25
15114865 Female Luer x 1/4 to 28 in. UNF pezzi 25
15116116 Female x Female x Female pezzi 25
15102954 Male LL to 1/4 to 28 UNF Thread pezzi 25
15395631 Male LL to 10 to 32 Thread pezzi 25
15329612 Male x Female x Female 25 confezioni
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Codice prodotto. 15305641 Fornitore Masterflex™ N. del fornitore 4550826
  • ONLINE EXCLUSIVE PRICE
    45.10€
    42.38€ / pezzi 25
    Risparmia 2.72€
    6% Sconto
Visualizza tutte le promozioni
Spedizione stimata: 09-09-2026
Aggiungi al carrello
Aggiungi al carrello
Featuring a variety of fitting options, these Luer fittings are ideal for many applications including syringes or small tubing. Cole-Parmer™ Polypropylene Luer Fitting are available with standard, wide, and low-profile barbs.
  • Three barb styles: standard, wide-bore, and low-profile.
    • Wide-bore barbs offer enhanced fluid flow through the fitting and are usable with any flexible tubing.
    • Low-profile barbs are ideal for use with semi-rigid tubing, such as nylon, polyethylene, or polypropylene.
  • Manufactured in compliance with ISO 594-1 standards, these fittings are cleaned, inspected and fully traceable.
  • All parts meet tight dimensional tolerances for increased reliability and process purity.
  • Choose from a range of sizes, 1/16in. to 1/4in., with a variety of connection options.
  • Pack of 25
Specifica
Certificazione/Conformità ISO 594-1 standards
Materiale Polipropilene
Da utilizzare con Female Luer x Female Luer, Elbow
Quantità confezione 25 Pack
Product Type Luer Fitting