Descrizione
- Manufactured of animal-free natural polypropylene
- All parts meet tight dimensional tolerances for increased reliability and process purity
- Choose from a range of sizes, with a variety of connection options
- Good chemical resistance. Max temperature 87°C (190°F)
- Sterilize by ethylene oxide or gamma irradiation (not recommended for autoclaving)
- All female ports may be used with either slip or luer lock connections
- Plugs and caps are also available
Specifica
Specifica
| Materiale | Polypropylene |
| Da utilizzare con | Fits Male Luer Lock to 1/8 in. I.D. |
| Da utilizzare con (applicazione) | Ideal for a variety of applications, including syringes or small tubing |
| Quantità confezione | 25 Pack |
| Product Type | Luer Adapter |