Learn More
Merck Millipore Polypropylene Microfil Funnel and Filter
Convenient, easy to use Microfil™ membrane funnel and filteration system
Marca:  Merck Millipore MIHCWG100
Dettagli aggiuntivi : Peso : 1.00000kg
Codice prodotto. 12841556
  • 236.00€ / pezzi 150
Spedizione stimata
10-03-2021
Aggiungi al carrello
Descrizione
Descrizione
  • Convenient, easy to use system for well accepted membrane filtration method conforms to international standards: CEN, Standard Methods, WHO
  • Disposable, stackable, light, recyclable plastic Microfil™ funnels eliminate autoclaving and minimise waste
  • 'Push fit' Microfil™ funnels tightly seal to manifold supports without clamps or O-rings assuring leak-free operation and uniform micro-organism recoveries
  • Novel device lifts membrane after filtration, for easy removal with forceps
  • Unique Microfil™ support filters faster than all-glass filter apparatus
  • 100mL or 250mL funnels available with convenient accessory dispensers
  • Choose conventional sterile membranes or novel EZ-Pak™ membrane cartridges. Select manifold-mounted filtration supports for laboratory use or stand-alone on-site filtration support.
Specifica
Specifica
White
Chlorinated Effluents, Clinical Lab, Food and Beverage, Industrial QC , Environmental Analysis, Laboratory Filtration
150/Pack
0.7μm
100mL
47mm
Polypropylene
Microfil Funnel and Filter