Learn More
Brady™ Overlaminate for Padlock Labels
Product Code. 15603617 Shop All Brady Products
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 40
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15603617 Clear Pack of 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15603617 Supplier Brady™ Supplier No. 065508
  • ONLINE EXCLUSIVE PRICE
    26.20€
    24.14€ / pezzi 40
    Risparmia 2.06€
    7% Sconto
Spedizione stimata: 29-12-2026
Polyester Overlaminate
  • Contains Permanent Acrylic adhesive
Specifications
Product Type Padlock Labels
Alternate Part Number B674 1.0X6.0 LOCKOUT OVERLAM 40 PCS/PK
Color Clear
Description Overlaminate for Padlock Labels
Quantity 1 Ea.