Learn More
VITLAB™ Polypropylene Pipette Stand
Description
- Polypropylene
- 94 bore holes of different diameters
- Easy assembly
Specifications
Specifica
| Material | Polypropylene |
| Positions | 94 Any Size |
| Quantity | 1 Each |
| Diameter (Metric) Cavity | 230 mm |
| Product Type | Pipette Stand |
| Height (Metric) | 470 mm |