Learn More
Greiner Bio-One Polystyrene Conical Bottom Analyzer Cups
Codice prodotto. 15965311
Cambia vista
Click to view available options
Volume (sistema metrico):
1.5 mL
4 mL
Da utilizzare con (apparecchiatura):
Technicon Analyzer
Dimensione della confezione:
pezzi 1000
pezzi 5000
pezzi 6000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Volume (sistema metrico) Da utilizzare con (apparecchiatura) unitSize
15965311 1.5 mL Technicon Analyzer pezzi 6000
10660742 4 mL Technicon Analyzer pezzi 1000
15975311 4 mL Technicon Analyzer pezzi 5000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Codice prodotto. 15965311 Fornitore Greiner Bio-One N. del fornitore 620101
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
Greiner Bio-One Cell 1.5mL Polystyrene Conical Bottom Analyzer Cups suitable for use with analytical systems, such as Technicon, Gemsaec, Centrifichem, Hitachi, TOA and Coulter analyzers.
  • Material: Polystyrene
  • Conical bottom
  • With support skirt
  • For single use only
TRUSTED_SUSTAINABILITY
Specifica
Da utilizzare con (apparecchiatura) Technicon Analyzer
Forma Conical
Sterilità Non-sterile
Colore Clear
Materiale Polystyrene
Product Type Analyzer Cup
Volume (sistema metrico) 1.5 mL
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.