Learn More
Macherey-Nagel™ Polystyrene Divinylbenzene Chromabond LV-HR-X
Marca: Macherey-Nagel™ 732132
Dettagli aggiuntivi : Peso : 0.50000kg
Specifica
200 mg | |
45 or 85 μm | |
1 to 14 | |
30 Pack |
Chromabond LV-HR-X | |
Polystyrene Divinylbenzene | |
55 - 60 Å |