Learn More
LABPLAS Polyurethane Sani-Stick with Neutralizing Buffer
Descrizione
- Polyurethane Sani-Stick
- Saturated with neutralizing buffer
- Dimensions: 160 x 229 mm
- Holds: 600 mL
- Option to include gloves with kit
Specifica
Specifica
| Product Type | Hygiene Sponge Sampling Kit |
| Tipo di media | Neutralizers |
| Sterilità | Sterile |
| Include | None |