Learn More
Tocris Bioscience™ Pomalidomide
Cereblon binder; also TNF-α inhibitor and antiangiogenic
Marca: Tocris Bioscience™ 6302/10
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Pomalidomide is a potent TNF-α inhibitor (IC50 = 13 nM). Also potently inhibits IL-2 (EC50 = 8 nM). Thalidomide derivative. Binds cereblon and inhibits its ubiquitination. Exhibits antiproliferative effects in a Namalwa lymphoma cell line. Antiangiogenic. Promotes degradation of transcription factor SALL4.Specifica
| 4-Amino-2-(2,6-dioxo-3-piperidinyl)-1 H-isoindole-1,3(2 H)-dione | |
| 19171-19-8 | |
| >98% | |
| 10 mg |